Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH991805A7

AstaTech, Inc.

TERT-BUTYL 3-HYDROXY-6-AZABICYCLO[3.2.1]OCTANE-6-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
227.3
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

tert-butyl (1R,5S)-3-hydroxy-6-azabicyclo[3.2.1]octane-6-carboxylate

Inchi Code

1S/C12H21NO3/c1-12(2,3)16-11(15)13-7-8-4-9(13)6-10(14)5-8/h8-10,14H,4-7H2,1-3H3

InChI key

UYQWTUFQGNHJIK-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=165800

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W19579


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service