Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH98B84D90

AstaTech, Inc.

2-ETHOXY-6-NITROACRIDIN-9-AMINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
283.29
MDL number:

Iupac Name

2-ethoxy-6-nitro-9-acridinamine

Inchi Code

1S/C15H13N3O3/c1-2-21-10-4-6-13-12(8-10)15(16)11-5-3-9(18(19)20)7-14(11)17-13/h3-8H,2H2,1H3,(H2,16,17)

InChI key

NDKYICANXPTKKC-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Light-red to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=246324

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=E83758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service