Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH97E805B2

AstaTech, Inc.

6-CHLORO-1-METHYL-1H-PYRAZOLO[3,4-B]PYRAZINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
168.59
MDL number:

Iupac Name

6-chloro-1-methyl-1H-pyrazolo[3,4-b]pyrazine

Inchi Code

1S/C6H5ClN4/c1-11-6-4(2-9-11)8-3-5(7)10-6/h2-3H,1H3

InChI key

RDLJOWUJZXXHOH-UHFFFAOYSA-N

Purity

95%

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=195300

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D78495


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service