Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATEH97B583CB

AstaTech, Inc.

2-AMINO-4-FLUORO-1,3-BENZOTHIAZOLE-6-CARBONITRILE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.2
MDL number:

Iupac Name

2-amino-4-fluorobenzo[d]thiazole-6-carbonitrile

Inchi Code

1S/C8H4FN3S/c9-5-1-4(3-10)2-6-7(5)12-8(11)13-6/h1-2H,(H2,11,12)

InChI key

HQJWVMZREMMKQY-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Sticky Oil to Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=172607

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=N17016


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service