Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE638639603

AstaTech, Inc.

2-CHLORO-5-(CYCLOPENTYLOXY)PYRIMIDINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
198.65
MDL number:

Iupac Name

2-chloro-5-(cyclopentyloxy)pyrimidine

Inchi Code

1S/C9H11ClN2O/c10-9-11-5-8(6-12-9)13-7-3-1-2-4-7/h5-7H,1-4H2

InChI key

MANHSBQXHPLZSR-UHFFFAOYSA-N

Purity

95%

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=100582

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=92044


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service