Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE514322682

AstaTech, Inc.

2-((1E,3E)-3-(3-(6-((2,5-DIOXOPYRROLIDIN-1-YL)OXY)-6-OXOHEXYL)BENZO[D]OXAZOL-2(3H)-YLIDENE)PROP-1-EN-1-YL)-3-ETHYLBENZO[D]OXAZOL-3-IUM IODIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
643.48
MDL number:

Iupac Name

2-((1E,3E)-3-(3-(6-((2,5-dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)benzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)-3-ethylbenzo[d]oxazol-3-ium iodide

Inchi Code

1S/C29H30N3O6.HI/c1-2-30-21-11-5-7-13-23(21)36-27(30)15-10-16-28-31(22-12-6-8-14-24(22)37-28)20-9-3-4-17-29(35)38-32-25(33)18-19-26(32)34;/h5-8,10-16H,2-4,9,17-20H2,1H3;1H/q+1;/p-1

InChI key

RQYPAAVMQSMNBY-UHFFFAOYSA-M

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Light-red to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=87175

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F14630


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service