Skip to Content
MilliporeSigma
All Photos(1)

Documents

ATE410297192

AstaTech, Inc.

2-METHOXY-4,6-DIMETHYLNICOTINIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
181.19
MDL number:

Iupac Name

2-methoxy-4,6-dimethylnicotinic acid

Inchi Code

1S/C9H11NO3/c1-5-4-6(2)10-8(13-3)7(5)9(11)12/h4H,1-3H3,(H,11,12)

InChI key

OILYJALBGQMVGR-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=21504

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=84032


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service