Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATE316039900

AstaTech, Inc.

2-(PYRROLIDIN-2-YL)ETHANOL HCL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
151.64
MDL number:

Iupac Name

2-(2-pyrrolidinyl)ethanol hydrochloride

Inchi Code

1S/C6H13NO.ClH/c8-5-3-6-2-1-4-7-6;/h6-8H,1-5H2;1H

InChI key

VLCKHADTUGEQCH-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Yellow to Brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=7079

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=62964


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service