Skip to Content
MilliporeSigma
All Photos(1)

Documents

APOH316CDED1

Apollo Scientific Ltd

2-Bromo-1-chloro-3-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
209.44
MDL number:

Iupac Name

2-bromo-1-chloro-3-fluorobenzene

Inchi Code

1S/C6H3BrClF/c7-6-4(8)2-1-3-5(6)9/h1-3H

InChI key

GBESIUPWXGQOFP-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/2-bromo-1-chloro-3-fluorobenzene

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-bromo-1-chloro-3-fluorobenzene


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service