Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH086AB655

Apollo Scientific Ltd

2,3-Difluoro-4-methylphenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
249.05
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

2-bromo-1-(2,3-difluoro-4-methylphenyl)ethanone

Inchi Code

1S/C9H7BrF2O/c1-5-2-3-6(7(13)4-10)9(12)8(5)11/h2-3H,4H2,1H3

InChI key

ISFMNZDYUUEHJC-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/23-difluoro-4-methylphenacyl-bromide

Synonym

2-Bromo-1-(2,3-difluoro-4-methylphenyl)ethan-1-one, 2-Bromo-2',3'-difluoro-4'-methylacetophenone

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/23-difluoro-4-methylphenacyl-bromide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service