Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH086AAE65

Apollo Scientific Ltd

5-Fluoro-6-(trifluoromethyl)nicotinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
190.1
MDL number:

Iupac Name

5-fluoro-6-(trifluoromethyl)nicotinonitrile

Inchi Code

1S/C7H2F4N2/c8-5-1-4(2-12)3-13-6(5)7(9,10)11/h1,3H

InChI key

HSJJRYVCFVSJAZ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Oil

Product Link

https://store.apolloscientific.co.uk/product/5-fluoro-6-trifluoromethylnicotinonitrile

Synonym

5-Fluoro-6-(trifluoromethyl)pyridine-3-nitrile

Pictograms

Skull and crossbones

Signal Word

Danger

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-fluoro-6-trifluoromethylnicotinonitrile


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service