Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APOH04239DE1

Apollo Scientific Ltd

2-Chloro-6-(trifluoromethoxy)benzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
289.48
MDL number:

Iupac Name

2-(bromomethyl)-1-chloro-3-(trifluoromethoxy)benzene

Inchi Code

1S/C8H5BrClF3O/c9-4-5-6(10)2-1-3-7(5)14-8(11,12)13/h1-3H,4H2

InChI key

COHYTMOYGPWHHN-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Oil

Product Link

https://store.apolloscientific.co.uk/product/2-chloro-6-trifluoromethoxybenzyl-bromide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-chloro-6-trifluoromethoxybenzyl-bromide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service