Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO959408682

Apollo Scientific Ltd

Ethyl 2,2-difluorohexanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
180.19
MDL number:

Iupac Name

ethyl 2,2-difluorohexanoate

Inchi Code

1S/C8H14F2O2/c1-3-5-6-8(9,10)7(11)12-4-2/h3-6H2,1-2H3

InChI key

XTUTXYBGTIFZFQ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Clear

Product Link

https://store.apolloscientific.co.uk/product/ethyl-22-difluorohexanoate

Synonym

Ethyl 2,2-difluorocaproate

Flash Point

83-85/15mm

Pictograms

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/ethyl-22-difluorohexanoate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service