Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO458955676

Apollo Scientific Ltd

tert-Butyl 3-aminopropanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
145.2
MDL number:

mp

64

Iupac Name

tert-butyl 3-aminopropanoate

Inchi Code

1S/C7H15NO2/c1-7(2,3)10-6(9)4-5-8/h4-5,8H2,1-3H3

InChI key

ZJXHVYSDMUKUCA-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/tert-butyl-3-aminopropanoate

Flash Point

192.3

Melting Point

64

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/tert-butyl-3-aminopropanoate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service