Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456796309

Apollo Scientific Ltd

1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
337.89
MDL number:

mp

none

density

2.003

Iupac Name

1,5-dibromo-1,1,3,3,5,5-hexafluoropentane

Inchi Code

1S/C5H4Br2F6/c6-4(10,11)1-3(8,9)2-5(7,12)13/h1-2H2

InChI key

XYMBGGTVDWPIBA-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/15-dibromo-113355-hexafluoropentane

Flash Point

54.2/14mm

Melting Point

none

density

2.003

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/15-dibromo-113355-hexafluoropentane


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service