Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455838245

Apollo Scientific Ltd

2,4,5-Trifluorophenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
171.12
MDL number:

density

1.35

Iupac Name

(2,4,5-trifluorophenyl)acetonitrile

Inchi Code

1S/C8H4F3N/c9-6-4-8(11)7(10)3-5(6)1-2-12/h3-4H,1H2

InChI key

JTYBTJVFXUKNKW-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/245-trifluorophenylacetonitrile

Synonym

2,4,5-Trifluorobenzyl cyanide

Flash Point

84/1mm

density

1.35

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/245-trifluorophenylacetonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service