Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455836640

Apollo Scientific Ltd

4-Bromo-2-(trifluoromethyl)quinoline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
276.06
MDL number:

Iupac Name

4-bromo-2-(trifluoromethyl)quinoline

Inchi Code

1S/C10H5BrF3N/c11-7-5-9(10(12,13)14)15-8-4-2-1-3-6(7)8/h1-5H

InChI key

JRTGGNDDSKKPQE-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

96%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethylquinoline

Synonym

4-Bromo-2-(trifluoromethyl)-1-azanaphthalene

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethylquinoline


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service