Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455835608

Apollo Scientific Ltd

(4,5-Dichloro-1H-imidazol-1-yl)methyl [4-(trifluoromethoxy)phenyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
370.11
MDL number:

Iupac Name

(4,5-dichloro-1H-imidazol-1-yl)methyl 4-(trifluoromethoxy)phenylcarbamate

Inchi Code

1S/C12H8Cl2F3N3O3/c13-9-10(14)20(5-18-9)6-22-11(21)19-7-1-3-8(4-2-7)23-12(15,16)17/h1-5H,6H2,(H,19,21)

InChI key

QLURORHHEYVXSA-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/45-dichloro-1h-imidazol-1-ylmethyl-4-trifluoromethoxyphenylcarbamate

Synonym

4,5-Dichloro-1-[({[4-(trifluoromethoxy)phenyl]carbamoyl}oxy)methyl]-1H-imidazole

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/45-dichloro-1h-imidazol-1-ylmethyl-4-trifluoromethoxyphenylcarbamate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service