Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455834325

Apollo Scientific Ltd

2,4,5-Trifluorophenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
253.02
MDL number:

bp

58-60

Iupac Name

2-bromo-1-(2,4,5-trifluorophenyl)ethanone

Inchi Code

1S/C8H4BrF3O/c9-3-8(13)4-1-6(11)7(12)2-5(4)10/h1-2H,3H2

InChI key

FUXKXNLPKBSNNE-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/245-trifluorophenacyl-bromide

Synonym

2-Bromo-1-(2,4,5-trifluorophenyl)ethan-1-one, 2-Bromo-2',4',5'-trifluoroacetophenone

Boiling point

58-60

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/245-trifluorophenacyl-bromide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service