Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455833609

Apollo Scientific Ltd

3-Bromo-5-chloro-1,2,4-thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
199.46
MDL number:

bp

24-25

Iupac Name

3-bromo-5-chloro-1,2,4-thiadiazole

Inchi Code

1S/C2BrClN2S/c3-1-5-2(4)7-6-1

InChI key

CXUWGEWQRCXJDC-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Low Melt

Product Link

https://store.apolloscientific.co.uk/product/3-bromo-5-chloro-124-thiadiazole

Flash Point

192-194

Boiling point

24-25

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-bromo-5-chloro-124-thiadiazole


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service