Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455833137

Apollo Scientific Ltd

4-(5-Chlorothien-2-yl)-1,3-thiazol-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
216.71
MDL number:

bp

129-130

Iupac Name

4-(5-chloro-2-thienyl)-1,3-thiazol-2-amine

Inchi Code

1S/C7H5ClN2S2/c8-6-2-1-5(12-6)4-3-11-7(9)10-4/h1-3H,(H2,9,10)

InChI key

KZOUOGWZDXYEDI-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-5-chlorothien-2-yl-13-thiazol-2-amine

Boiling point

129-130

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-5-chlorothien-2-yl-13-thiazol-2-amine


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service