Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455828690

Apollo Scientific Ltd

Bicyclo[2.2.1]heptane-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
140.18
MDL number:

bp

40

mp

>100

Iupac Name

bicyclo[2.2.1]heptane-2-carboxylic acid

Inchi Code

1S/C8H12O2/c9-8(10)7-4-5-1-2-6(7)3-5/h5-7H,1-4H2,(H,9,10)

InChI key

JESWDXIHOJGWBP-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Fused

Product Link

https://store.apolloscientific.co.uk/product/bicyclo221heptane-2-carboxylic-acid

Flash Point

134-136/16mm

Boiling point

40

Melting Point

>100

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/bicyclo221heptane-2-carboxylic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service