Skip to Content
MilliporeSigma

APO455828562

Apollo Scientific Ltd

1,4-Dihydropyrazine-2,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
112.09
MDL number:

Pricing and availability is not currently available.

Iupac Name

1,4-dihydro-2,3-pyrazinedione

Inchi Code

1S/C4H4N2O2/c7-3-4(8)6-2-1-5-3/h1-2H,(H,5,7)(H,6,8)

InChI key

CIGLZLCKRXOFQU-UHFFFAOYSA-N

storage temp.

Store at -20C

Country of Origin

GB

Shipping Temperature

Shipped with Ice Packs

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/14-dihydropyrazine-23-dione

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/14-dihydropyrazine-23-dione


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service