Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO428498607

Apollo Scientific Ltd

2,3-Difluoro-4-methoxybenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
174.15
MDL number:

Iupac Name

(2,3-difluoro-4-methoxyphenyl)methanol

Inchi Code

1S/C8H8F2O2/c1-12-6-3-2-5(4-11)7(9)8(6)10/h2-3,11H,4H2,1H3

InChI key

SJKZXUYOJRZFKG-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/23-difluoro-4-methoxybenzyl-alcohol

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/23-difluoro-4-methoxybenzyl-alcohol


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service