Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AOBH961DBEF0

AOBChem

Methyl 2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
280.22
MDL number:

Iupac Name

methyl 2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzoate

Inchi Code

1S/C12H12F4O3/c1-6(2)19-9-5-7(12(14,15)16)4-8(10(9)13)11(17)18-3/h4-6H,1-3H3

InChI key

FXXHREJZDPAJLW-UHFFFAOYSA-N

Purity

97

Country of Origin

CN

Physical Form

Liquid

Product Link

https://aobchem.com/methyl-2-fluoro-3-isopropoxy-5-trifluoromethyl-benzoate.html

MSDS

https://aobchem.com/pub/media/files/sds/38306.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service