Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH2D6F029C

Ambeed, Inc.

6-Methyl-2,4-dihydroxyquinoline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
175.19
MDL number:

Iupac Name

6-methyl-2,4(1H,3H)-quinolinedione

Inchi Code

1S/C10H9NO2/c1-6-2-3-8-7(4-6)9(12)5-10(13)11-8/h2-5H,1H3,(H2,11,12,13)

InChI key

OXSZQTDCCMODLE-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/1677-44-7.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/1677-44-7.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service