Key Documents
Z222852
Quick-connect hose coupling
Coupling, for tubing i.d., 3/8 in.
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
feature
for tubing i.d. 3/8 in.
General description
Nickel-plated brass coupling and nipple. Simple push-to-connect operation. Suitable for both pressure and vacuum applications. Max. pressure: 508 psi, min. burst pressure: 2,030 psi. Temp. range: -40 to 100 °C (-40 to 212 °F).
Choose from one of the most recent versions:
Certificates of Analysis (COA)