Skip to Content
MilliporeSigma
All Photos(1)

Documents

W6977

Methacetin-(methoxy-13C)

API for Clinical Studies

Synonym(s):

N-(4-Methoxy-13C-phenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4O13CH3
CAS Number:
Molecular Weight:
166.18
MDL number:
UNSPSC Code:
12142200

grade

cGMP

isotopic purity

≥99 atom % 13C

mp

128-130 °C (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

CC(=O)Nc1ccc(O[13CH3])cc1

InChI

1S/C9H11NO2/c1-7(11)10-8-3-5-9(12-2)6-4-8/h3-6H,1-2H3,(H,10,11)/i2+1

InChI key

XVAIDCNLVLTVFM-VQEHIDDOSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product is available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service