Skip to Content
MilliporeSigma
All Photos(1)

Documents

W6586

L-Phenyl-d5-alanine

greener alternative

API for Clinical Studies, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
170.22
MDL number:
UNSPSC Code:
12352116

grade

cGMP

isotopic purity

98 atom % D

Assay

99% (CP)

greener alternative product score

old score: 47
new score: 32
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

270-275 °C (dec.) (lit.)

greener alternative category

mass shift

M+5

storage temp.

15-25°C

SMILES string

[H]C(N)(Cc1c([2H])c([2H])c([2H])c([2H])c1[2H])C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2D,3D,4D,5D

InChI key

COLNVLDHVKWLRT-HRVDQBBZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service