Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W382701

Sigma-Aldrich

Isopropyl disulfide

≥96%, FG

Synonym(s):

Diisopropyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2S2
CAS Number:
Molecular Weight:
150.31
FEMA Number:
3827
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.109
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥96%

refractive index

n20/D 1.4906 (lit.)

bp

175-176 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; alliaceous; sulfurous

SMILES string

CC(C)SSC(C)C

InChI

1S/C6H14S2/c1-5(2)7-8-6(3)4/h5-6H,1-4H3

InChI key

LZAZXBXPKRULLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl disulfide is a volatile sulfur compound that is reported to occur in Allium species.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidative activity of sulfur?containing compounds in Allium species for human LDL oxidation in vitro.
Nishimura H, et al.
Biofactors, 21(1?4), 277-280 (2004)
Bofan Yu et al.
Journal of chromatography. A, 1356, 221-229 (2014-07-07)
For the expanded application area, fast trace analysis of certain high boiling point (i.e., 150-250 °C) volatile organic compounds (HVOCs) in water, a large volume-direct aqueous injection-gas chromatography (LV-DAI-GC) method was optimized for the following parameters: packed sorbent for sample

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service