Skip to Content
MilliporeSigma
All Photos(1)

Documents

W316806

Sigma-Aldrich

3,4-Hexanedione

≥96%

Synonym(s):

Dipropionyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CH2COCOCH2CH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
1700837
MDL number:
Flavis number:
7.077

Assay

≥96%

refractive index

n20/D 1.41 (lit.)

bp

131 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)C(=O)CC

InChI

1S/C6H10O2/c1-3-5(7)6(8)4-2/h3-4H2,1-2H3

InChI key

KVFQMAZOBTXCAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Butter, toasted, nutty, almond

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service