Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W269440

Sigma-Aldrich

Methyl isobutyrate

natural, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOOCH3
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2694
Beilstein:
1740720
EC Number:
Council of Europe no.:
287
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.412
NACRES:
NA.21

grade

Halal
natural

reg. compliance

FDA 21 CFR 117

Assay

98%

refractive index

n20/D 1.384 (lit.)

bp

90 °C (lit.)

density

0.891 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; fruity; floral; sweet

SMILES string

COC(=O)C(C)C

InChI

1S/C5H10O2/c1-4(2)5(6)7-3/h4H,1-3H3

InChI key

BHIWKHZACMWKOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service