Skip to Content
MilliporeSigma
All Photos(1)

Documents

W224618

Sigma-Aldrich

Carvacryl ethyl ether

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
FEMA Number:
2246
EC Number:
Council of Europe no.:
11840
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.038

grade

Kosher

Assay

≥98%

refractive index

n20/D 1.4965 (lit.)

bp

229 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

CCOc1cc(ccc1C)C(C)C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service