Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W203300

Sigma-Aldrich

Allyl-α-ionone

≥65%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O
CAS Number:
Molecular Weight:
232.36
FEMA Number:
2033
EC Number:
Council of Europe no.:
2040
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.061

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 172.515

concentration

≥65%

impurities

13% allyl β-ionone.

refractive index

n20/D 1.505 (lit.)

bp

102-104 °C/0.2 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; fruity; floral; spicy; woody

SMILES string

CC1=CCCC(C)(C)C1\C=C\C(=O)CCC=C

InChI

1S/C16H24O/c1-5-6-9-14(17)10-11-15-13(2)8-7-12-16(15,3)4/h5,8,10-11,15H,1,6-7,9,12H2,2-4H3/b11-10+

InChI key

FXCYGAGBPZQRJE-ZHACJKMWSA-N

Packaging

Packaged in glass bottles

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service