Documents
SYNF222500
Synthware four neck round bottom flask with angled side necks
capacity 500 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Synonym(s):
Synthware F222500, 4-neck reaction flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS29/42
side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F222500
capacity
500 mL
General description
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.