Skip to Content
Merck
All Photos(1)

Key Documents

R658588

7-((2,6-DICHLOROBENZYL)OXY)-8-METHYL-4-PHENYL-2H-CHROMEN-2-ONE

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C23H16Cl2O3
CAS Number:
Molecular Weight:
411.288
MDL number:

Pricing and availability is not currently available.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHR3258PHR3106PHR1798
application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

agency

traceable to USP 1611900

agency

traceable to USP 1695004

agency

traceable to USP 1561008

agency

traceable to BP 212, traceable to Ph. Eur. L0380000, traceable to USP 1359302

API family

selegiline

API family

tripelennamine

API family

prilocaine

API family

levamisole

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

packaging

pkg of 500 mg

packaging

pkg of 500 mg

packaging

pkg of 250 mg

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service