Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6808

Sigma-Aldrich

Mesitaldehyde

contains ~0.1% Hydroquinone as stabilizer, 98%

Synonym(s):

2,4,6-Trimethylbenzaldehyde, 2-Formyl-1,3,5-trimethylbenzene, 2-Formylmesitylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CHO
CAS Number:
Molecular Weight:
148.20
Beilstein:
1364114
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

~0.1% Hydroquinone as stabilizer

refractive index

n20/D 1.553 (lit.)

bp

237 °C (lit.)

mp

10-12 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c(C=O)c(C)c1

InChI

1S/C10H12O/c1-7-4-8(2)10(6-11)9(3)5-7/h4-6H,1-3H3

InChI key

HIKRJHFHGKZKRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service