Skip to Content
MilliporeSigma
All Photos(1)

Documents

H6827

Sigma-Aldrich

2-Hexadecanol

99%

Synonym(s):

Methyl tetradecyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH(OH)CH3
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748421
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

43-46 °C (lit.)

SMILES string

CCCCCCCCCCCCCCC(C)O

InChI

1S/C16H34O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16(2)17/h16-17H,3-15H2,1-2H3

InChI key

FVDRFBGMOWJEOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumikazu Takahashi et al.
FEMS yeast research, 19(2) (2019-02-13)
The yeast Starmerella bombicola NBRC10243 is an excellent producer of sophorolipids, which are among the most useful biosurfactants. The primary alcoholic metabolic pathway of S. bombicola has been elucidated using alcohol oxidase FAO1, but the secondary alcohol metabolic pathway remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service