Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF83693885

Waspaloy - heat resisting alloy, Ni58Cr19Co14MoTiAlFe

foil, 15mm disks, 0.025mm thickness, as rolled

Synonym(s):

NI150250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (disks - as rolled)

manufacturer/tradename

Goodfellow 836-938-85

size × thickness

15 mm × 0.025 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service