Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAF183090250

SP Bel-Art® Weighted Beaker/Flask Holder with Vikem Vinyl Coating

for use with 250 mL beakers

Synonym(s):

beaker holder, flask holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F18309-0250

compatibility

for use with 250 mL beakers

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service