Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A28708

Sigma-Aldrich

Allyl alcohol

99%

Synonym(s):

2-Propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OH
CAS Number:
Molecular Weight:
58.08
Beilstein:
605307
EC Number:
MDL number:
NACRES:
NA.24

vapor density

2 (vs air)

vapor pressure

23.8 mmHg ( 25 °C)

Assay

99%

autoignition temp.

712 °F

expl. lim.

18 %

refractive index

n20/D 1.412 (lit.)

bp

96-98 °C (lit.)

mp

−129 °C (lit.)

density

0.854 g/mL at 25 °C (lit.)

SMILES string

OCC=C

InChI

1S/C3H6O/c1-2-3-4/h2,4H,1,3H2

InChI key

XXROGKLTLUQVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to induce a mouse model of liver damage that has been used to study the mechanisms of hepatotoxicity and hepatic stem cell-mediated repair.

Packaging

18L packaged in Kilo-Lab™ metal cylinders. Cylinder-outlet valve or transfer line needed to dispense material. Cylinders require deposit

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taku Chiba et al.
Bioorganic & medicinal chemistry letters, 17(9), 2487-2491 (2007-03-03)
A series of sugar derivatives (7-14) were synthesized from stachyose, a sugar compound of Stachys sieboldi Miq, and evaluated for antibacterial activity against Mycobacterium tuberculosis, Mycobacterium avium, and Staphylococcus aureus, and their structure-activity relationships were studied. The results showed that
Takeki Uehara et al.
Molecular nutrition & food research, 54(2), 218-227 (2009-12-31)
Biotechnology advances have provided novel methods for the risk assessment of chemicals. The application of microarray technologies to toxicology, known as toxicogenomics, is becoming an accepted approach for identifying chemicals with potential safety problems. Gene expression profiling is expected to
Sean Ekins et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(12), 2302-2308 (2010-09-17)
Drug-induced liver injury (DILI) is one of the most important reasons for drug development failure at both preapproval and postapproval stages. There has been increased interest in developing predictive in vivo, in vitro, and in silico models to identify compounds
Nigel Greene et al.
Chemical research in toxicology, 23(7), 1215-1222 (2010-06-18)
Drug-induced liver injury is a major issue of concern and has led to the withdrawal of a significant number of marketed drugs. An understanding of structure-activity relationships (SARs) of chemicals can make a significant contribution to the identification of potential
Zhichao Liu et al.
PLoS computational biology, 7(12), e1002310-e1002310 (2011-12-24)
Drug-induced liver injury (DILI) is a significant concern in drug development due to the poor concordance between preclinical and clinical findings of liver toxicity. We hypothesized that the DILI types (hepatotoxic side effects) seen in the clinic can be translated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service