Skip to Content
MilliporeSigma
All Photos(1)

Documents

94703

Sigma-Aldrich

(−)-(R)-Fmoc-4-thiazolidinecarboxylic acid

≥99.0% (HPLC)

Synonym(s):

(−)-Fmoc-L-thiaproline, Fmoc-Thi-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO4S
CAS Number:
Molecular Weight:
355.41
Beilstein:
9349047
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (HPLC)

optical activity

[α]20/D −71±3°, c = 1% in DMF

mp

153-158 °C

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H]1CSCN1C(=O)OCC2c3ccccc3-c4ccccc24

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service