Skip to Content
MilliporeSigma
All Photos(1)

Documents

931268

Sigma-Aldrich

[3-(2,4,6-Trimethylbenzyl)-1-(2,6-diisopropylphenyl)-4,5-dimethylimidazolylidene]gold chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36AuClN2
Molecular Weight:
621.01
UNSPSC Code:
12352101
NACRES:
NA.21

form

powder

Quality Level

Application

[3-(2,4,6-Trimethylbenzyl)-1-(2,6-diisopropylphenyl)-4,5-dimethylimidazolylidene]gold chloride is an NHC catalyst bearing a highly hindered and flexible unsymmetrical NHC ligand for hydrofunctionalization reactions. This catalyst can be used in hydroelement additions, element-element additions, carbo-element additions, hydrations, hydroaminations, hydroarylations, hyroalkoxylations, hydrocarboxylations, cyclizations, hydrofluorinations, as well as a variety of combined processes using tandem reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service