Skip to Content
MilliporeSigma
All Photos(2)

Documents

914037

Sigma-Aldrich

Tetrakis(4-bromophenyl) silane

≥96%

Synonym(s):

1,1′,1′′,1′′′-Silanetetrayltetrakis[4-bromobenzene], SiTPBr4, Tetra(4-bromophenyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16Br4Si
CAS Number:
Molecular Weight:
652.09
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

Assay

≥96%

form

powder

color

white

InChI

1S/C24H16Br4Si/c25-17-1-9-21(10-2-17)29(22-11-3-18(26)4-12-22,23-13-5-19(27)6-14-23)24-15-7-20(28)8-16-24/h1-16H

InChI key

VHVUXJWGYUSRIA-UHFFFAOYSA-N

General description

Tetrakis(bromophenyl) silane is an intermediate for emerging hole transport materials based on a silicon core. Materials containing this core have been shown to compete with SpiroMeOTAD for efficiency and stability in perovskite solar cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular design with silicon core: toward commercially available hole transport materials for high-performance planar p−i−n perovskite solar cells
Xue R, et al.
Journal of Material Chemistry A, 6, 404-404 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service