Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

909068

Sigma-Aldrich

Tritolylamine diacrylate

>98%

Synonym(s):

4,4′-di(acrylyloxypropyl)-4″-methyl-triphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33NO4
Molecular Weight:
483.60
UNSPSC Code:
12352116
NACRES:
NA.23

Assay

>98% (HPLC)
>98%

form

viscous liquid

contains

150-400 ppm MEHQ as stabilizer

color

yellow to brown

storage temp.

2-8°C

Application

Tritolylamine diacrylate is a polymerizable charge transport molecule. It can be used for long-life photoconductor coatings in laser printers.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service