Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900420

Sigma-Aldrich

Graphene nanoplatelets

15 μm particle size

Synonym(s):

xGnP® M-15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12352200

description

oxygen content: < 1%
residual acid content: <0.5 wt%

form

powder

surface area

120-150 m2/g

thickness

6-8 nm , average

particle size

15 μm

bulk density

0.03‑0.1 g/cm3

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

  • xGnP® graphene nanoplatelets are unique nanoparticles consisting of short stacks of graphene sheets having a platelet shape.
  • The unique size and platelet morphology of xGnP® graphene nanoplatelets makes these particles especially effective at providing barrier properties, while their pure graphitic composition makes them excellent electrical and thermal conductors. xGnP® graphene nanoplatelets can improve mechanical properties such as stiffness, strength, and surface hardness of the matrix material.
  • xGnP® graphene nanoplatelets are compatible with almost all polymers, and can be an active ingredient in inks or coatings as well as an excellent additive to plastics of all types. The unique manufacturing processes are non-oxidizing, so material has a pristine graphitic surface of sp2 carbon molecules that makes it especially suitable for applications requiring high electrical or thermal conductivity.
  • Grade M particles have an average thickness of approximately 6-8 nanometers and a typical surface area of 120 to 150 m2/g. Grade M is available with average particle diameters of 5, 15 or 25 microns.

Application

  • Ultracapacitor electrodes.
  • Anode materials for lithium-ion batteries.
  • Conductive additive for battery electrodes.
  • Electrically conductive inks.
  • Thermally conductive films and coatings.
  • Additive for lightweight composites.
  • Films or coatings for EMI shielding.
  • Substrate for chemical and biochemical sensors.
  • Barrier material for packaging.
  • Additive for super-strong concrete.
  • Additive for metal-matrix composites.

Other Notes

Graphene nanoplatelets have naturally occurring functional groups like ethers, carboxyls, or hydroxyls that can react with atmospheric humidity to form acids or other compounds. These functional groups are present on the edges of the particles and their wt% varies with particle size.

Legal Information

xGnP is a registered trademark of XG Sciences, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance dependence of thermosyphon on the functionalization approaches: An experimental study on thermo-physical properties of graphene nanoplatelet-based water nanofluids.
Amiri A, et al.
Energy Conversion and Management , 92, 322-330 (2015)
Mechanical properties of graphene nanoplatelet/epoxy composites.
King JA, et al.
Journal of Composite Materials, 49(6), 659-668 (2015)

Articles

Advances in scalable synthesis and processing of two-dimensional materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service