Skip to Content
MilliporeSigma
All Photos(1)

Documents

809195

Sigma-Aldrich

3-(Perfluoropyridin-4-yl)-3-propyl-1,5-dioxaspiro[5.5]undecane-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17F4NO4
CAS Number:
Molecular Weight:
375.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

storage temp.

2-8°C

SMILES string

FC1=C(F)N=C(F)C(F)=C1C2(CCC)C(OC3(CCCCC3)OC2=O)=O

InChI

1S/C17H17F4NO4/c1-2-6-17(9-10(18)12(20)22-13(21)11(9)19)14(23)25-16(26-15(17)24)7-4-3-5-8-16/h2-8H2,1H3

InChI key

OJBHZVGQJBBEDE-UHFFFAOYSA-N

General description

3-(Perfluoropyridin-4-yl)-3-propyl-1,5-dioxaspiro[5.5]undecane-2,4-dione is a substituted Meldrum′s acid derivative. It can be prepared by reacting pentafluoropyridine with 3-propyl-1,5-dioxaspiro[5.5]undecane-2,4-dione.

Application

This compound undergoes cycloreversion to form the fluoro(hetero)aryl ketene, which undergoes efficient coupling with nucleophiles and allows rapid incorporation of highly fluorinated α-fluoro(hetero)aryl acetic acid derivatives.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameera M Senaweera et al.
The Journal of organic chemistry, 79(21), 10466-10476 (2014-10-02)
This work describes the facile and mono-selective per- and polyfluoroarylation of Meldrum's acid to generate a versatile synthon for highly fluorinated α-phenyl acetic acid derivatives, which provide straightforward access to fluorinated building blocks. The reaction takes place quickly, and most

Related Content

Organofluorine chemistry is an essential part of drug discovery programs as well as agrochemical programs and even plays a major role in materials chemistry. Despite the undeniable importance of fluorinated organic molecules, our ability to synthesize these substrates is lacking - though arguably it is better than that of Nature. Consequently, methods that allow facile access to fluorinated molecules are important especially when they provide unique access to fluorinated chemical space.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service