Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

808474

Sigma-Aldrich

4arm-PEG10K

average Mn 10,000, norbornene

Synonym(s):

PEG-nb, PEG-norbornene, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C8H9O2(C2H4O)nCH2]4C
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

4-arm Poly(ethylene glycol) norbornene terminated, average Mn 10,000

form

powder

Quality Level

mol wt

average Mn 10,000

mp

43-48 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Multifunctional PEG for crosslinking, may be used to form hydrogels for biomaterial applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex A Aimetti et al.
Biomaterials, 30(30), 6048-6054 (2009-08-14)
Degradable hydrogels have been extensively used in biomedical applications such as drug delivery, and recent interest has grown in hydrogels that degrade in recognition of a cellular response. This contribution describes a poly(ethylene glycol) (PEG) hydrogel platform with human neutrophil

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service