Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

790516

Sigma-Aldrich

Lithium aluminum titanium phosphate

≥99.9% trace metals basis

Synonym(s):

Aluminum lithium titanium phosphate, LATP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li1.3Al0.3Ti1.7(PO4)3
CAS Number:
UNSPSC Code:
12352200

Assay

≥99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium aluminum titanium phosphate (LATP) is a ceramic lithium electrolyte for all solid state lithium battery. LATP is also used as the solid component for inorganic solid/organic liquid hybrid electrolyte for Li-ion batteries. The high lithium ion conductivity of LATP also makes them promising for use as Li+-conducting glass ceramic plate (LiGC plate) (separator membrane) in lithium air batteries.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

He; H.;
Electrochimica Acta, 67, 87-87 (2012)
Birke; P.;
Solid State Ionics, 118(1;2), 149-149 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service