Skip to Content
MilliporeSigma
All Photos(1)

Documents

771236

Sigma-Aldrich

2,6-Difluorophenylboronic acid MIDA ester

95%

Synonym(s):

2-(2,6-Difluorophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BF2NO4
Molecular Weight:
269.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

209-214 °C

SMILES string

FC1=C(C(F)=CC=C1)B2OC(CN(C)CC(O2)=O)=O

InChI

1S/C11H10BF2NO4/c1-15-5-9(16)18-12(19-10(17)6-15)11-7(13)3-2-4-8(11)14/h2-4H,5-6H2,1H3

InChI key

KARAINNDHRWYTL-UHFFFAOYSA-N

Application

Protected boronic acid with methylimindodiacetic acid (MIDA) that does not react under standard Suzuki-Miyuara coupling conditions, allowing for selective and iterative cross-coupling sequences. The MIDA boronate also provides prolonged shelf life and stability to typically unstable boronic acid derivatives.

MIDA Boronates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service